Merck
  • Time course of necrotizing hepatopancreatitis (NHP) in experimentally infected Litopenaeus vannamei and quantification of NHP-bacterium using real-time PCR.
Time course of necrotizing hepatopancreatitis (NHP) in experimentally infected Litopenaeus vannamei and quantification of NHP-bacterium using real-time PCR.
Diseases of aquatic organisms (2006-01-03)
Amanda G Vincent, Jeffrey M Lotz
ABSTRACT
Necrotizing hepatopancreatitis (NHP), a severe bacterial disease affecting penaeid shrimp aquaculture, is caused by a gram-negative, pleomorphic, intracellular alpha-proteobacterium referred to as the NHP-bacterium (NHPB). The time course of NHP was investigated in experimentally infected juveniles of Kona stock Litopenaeus vannamei. Susceptible animals were individually isolated in 41 of aerated artificial seawater at salinity 30 +/- 1 ppt and maintained in a water bath at 30 +/- 1 degree C for 60 d. A total of 120 individuals were exposed per os to a 0.05 g piece of NHPB-infected hepatopancreas and 100 controls were exposed to uninfected tissue. At intervals of 3, 6, 9, 16, 23, 30, 37, 44, and 53 d post-exposure, 6 shrimp exposed to NHPB-infected tissue and 4 controls were randomly removed from the experiment; hepatopancreas samples were processed for histological and molecular analysis, and feces were processed for molecular diagnosis of NHPB infection. NHPB was first detected in the hepatopancreas through histology at 6 d post-exposure. All control shrimp were diagnosed as NHPB negative. NHPB infections classified as stage I (scattering of hepatopancreatic tubules with adjacent epithelial cells containing NHPB) and stage II (numerous infected tubules with occasional hemocyte infiltration) were observed from 6 to 37 d post-exposure. All animals that experienced NHPB-induced mortality from 16 to 51 d post-exposure were at stage III (numerous necrotic tubules, dense hemocyte infiltration, and presence of granulomas). NHPB is capable of infecting all hepatopancreatic cell types including embryonic, resorptive, fibrillar and blister-like cells. The percent of hepatopancreatic tubules containing NHPB in epithelial cells increased over time, representing bacteria multiplication and spread. Real-time PCR allowed for quantification of NHPB in hepatopancreas and feces. Over the course of infection, NHPB was present at 10(3) to 10(7) copies mg(-1) of hepatopancreas and 10(1) to 10(5) copies mg(-1) of feces. Lethal infections contained 10(6) to 10(7) copies mg(-1) of hepatopancreas and 10(3) to 10(6) copies mg(-1) of feces.