Merck
  • Micropillar arrays as a high-throughput screening platform for therapeutics in multiple sclerosis.
Micropillar arrays as a high-throughput screening platform for therapeutics in multiple sclerosis.
Nature medicine (2014-07-07)
Feng Mei, Stephen P J Fancy, Yun-An A Shen, Jianqin Niu, Chao Zhao, Bryan Presley, Edna Miao, Seonok Lee, Sonia R Mayoral, Stephanie A Redmond, Ainhoa Etxeberria, Lan Xiao, Robin J M Franklin, Ari Green, Stephen L Hauser, Jonah R Chan
ABSTRACT
Functional screening for compounds that promote remyelination represents a major hurdle in the development of rational therapeutics for multiple sclerosis. Screening for remyelination is problematic, as myelination requires the presence of axons. Standard methods do not resolve cell-autonomous effects and are not suited for high-throughput formats. Here we describe a binary indicant for myelination using micropillar arrays (BIMA). Engineered with conical dimensions, micropillars permit resolution of the extent and length of membrane wrapping from a single two-dimensional image. Confocal imaging acquired from the base to the tip of the pillars allows for detection of concentric wrapping observed as 'rings' of myelin. The platform is formatted in 96-well plates, amenable to semiautomated random acquisition and automated detection and quantification. Upon screening 1,000 bioactive molecules, we identified a cluster of antimuscarinic compounds that enhance oligodendrocyte differentiation and remyelination. Our findings demonstrate a new high-throughput screening platform for potential regenerative therapeutics in multiple sclerosis.
Sigma-Aldrich
Anti-Myelin Basic Protein Antibody, a.a. 36-50, clone 14, culture supernatant, clone 14, Chemicon®
Pricing and availability is not currently available.