Merck
  • Astrocytic metabolic switch is a novel etiology for Cocaine and HIV-1 Tat-mediated neurotoxicity.
Astrocytic metabolic switch is a novel etiology for Cocaine and HIV-1 Tat-mediated neurotoxicity.
Cell death & disease (2018-03-20)
Kalimuthusamy Natarajaseenivasan, Bianca Cotto, Santhanam Shanmughapriya, Alyssa A Lombardi, Prasun K Datta, Muniswamy Madesh, John W Elrod, Kamel Khalili, Dianne Langford
ABSTRACT
Calcium (Ca2+) dynamics and oxidative signaling control mitochondrial bioenergetics in the central nervous system, where astrocytes are a major energy source for neurons. Cocaine use exacerbates HIV-associated neurocognitive disorders, but little is known about disruptions in astrocyte metabolism in this context. Our data show that the HIV protein Tat and cocaine induce a metabolic switch from glucose to fatty acid oxidation in astrocytes, thereby limiting lactate transport to neurons. Mechanistic analyses revealed increased Mitochondrial Ca2+ Uniporter (MCU)-mediated Ca2+ uptake in astrocytes exposed to Tat and cocaine due to oxidation of MCU. Since our data suggest that mitochondrial oxidation is dependent in part on MCU-mediated Ca2+ uptake, we targeted MCU to restore glycolysis in astrocytes to normalize extracellular lactate levels. Knocking down MCU in astrocytes prior to Tat and cocaine exposure prevented metabolic switching and protected neurons. These findings identify a novel molecular mechanism underlying neuropathogenesis in HIV and cocaine use.
Sigma-Aldrich
Lactate Assay Kit, sufficient for 100 colorimetric or fluorometric tests
Sigma-Aldrich
Lactate Dehydrogenase Activity Assay Kit, sufficient for 500 colorimetric tests
Sigma-Aldrich
Pyruvate Assay Kit, sufficient for 100 colorimetric or fluorometric tests