Merck
  • Point-of-service, quantitative analysis of ascorbic acid in aqueous humor for evaluating anterior globe integrity.
Point-of-service, quantitative analysis of ascorbic acid in aqueous humor for evaluating anterior globe integrity.
Scientific reports (2015-11-04)
Manas R Gartia, Santosh K Misra, Mao Ye, Aaron Schwartz-Duval, Lisa Plucinski, Xiangfei Zhou, David Kellner, Leanne T Labriola, Dipanjan Pan
ABSTRACT
Limited training, high cost, and low equipment mobility leads to inaccuracies in decision making and is concerning with serious ocular injuries such as suspected ruptured globe or post-operative infections. Here, we present a novel point-of-service (POS) quantitative ascorbic acid (AA) assay with use of the OcuCheck Biosensor. The present work describes the development and clinical testing of the paper-based biosensor that measures the changes in electrical resistance of the enzyme-plated interdigitated electrodes to quantify the level of AA present in ocular fluid. We have demonstrated the proof-of-concept of the biosensor testing 16 clinical samples collected from aqueous humor of patients undergoing therapeutic anterior chamber paracentesis. Comparing with gold standard colorimetric assay for AA concentration, OcuCheck showed accuracy of >80%, sensitivity of >88% and specificity of >71%. At present, there are no FDA-approved POS tests that can directly measures AA concentration levels in ocular fluid. We envisage that the device can be realized as a handheld, battery powered instrument that will have high impact on glaucoma care and point-of-care diagnostics of penetrating ocular globe injuries.
Sigma-Aldrich
Ascorbic Acid Assay Kit, sufficient for 100 colorimetric or fluorometric tests