Merck
  • Investigations concerning the COX/5-LOX inhibiting and hydroxyl radical scavenging potencies of novel 4,5-diaryl isoselenazoles.
Investigations concerning the COX/5-LOX inhibiting and hydroxyl radical scavenging potencies of novel 4,5-diaryl isoselenazoles.
European journal of medicinal chemistry (2007-11-03)
Michael Scholz, Holger K Ulbrich, Gerd Dannhardt
ABSTRACT
The aim of this study was to investigate 4,5-diaryl isoselenazoles as multiple target non-steroidal anti-inflammatory drugs (MTNSAIDs) which can intervene into the inflammatory processes via different mechanisms of action creating a new class of compounds. Here we describe the synthesis of COX/LOX inhibitors which additionally reduce the level of reactive oxygen species, such as hydroxyl radicals which are well known for supporting inflammation processes in Parkinson's disease, Alzheimer's disease and rheumatoid arthritis.
Sigma-Aldrich
Nordihydroguaiaretic acid, ≥90% (HPLC), from Larrea divaricata (creosote bush)
Sigma-Aldrich
Nordihydroguaiaretic acid, ≥97.0% (HPLC)
Supelco
(R)-(+)-α-Lipoic acid, analytical standard