Merck
  • Molecular mechanisms of beta2-adrenergic receptor function and regulation.
Molecular mechanisms of beta2-adrenergic receptor function and regulation.
Proceedings of the American Thoracic Society (2005-11-04)
Dennis W McGraw, Stephen B Liggett
ABSTRACT
It is now clear that the beta2-adrenergic receptor continuously oscillates between various conformations in the basal state, and that agonists act to stabilize one or more conformations. It is conceivable that synthetic agonists might be engineered to preferentially confine the receptor to certain conformations deemed clinically important while having a less stabilizing effect on unwanted conformations. In addition, studies of genetically engineered mice have revealed previously unrecognized cross-talk between the beta2-receptor and phospholipase C, such that removal of the primary dilating pathway results in downregulation of constrictive pathways and overactivity of the dilating pathway increases the contractile response. These results indicate a dynamic interaction between beta2-receptor activity and Gq-coupled receptors that constrict the airway. Potentially, then, during chronic beta-agonist therapy, expression of phospholipase C is increased, the functions of Gq-coupled constrictive receptors are enhanced, and there may be an increased tendency for clinical decompensation due to asthma and chronic obstructive pulmonary disease triggers. Antagonists to these receptors might be able to act synergistically with chronic beta-agonists to block the effect of phospholipase C. Alternatively, perhaps novel phospholipase C antagonists would provide the most efficacious approach to blocking the physiologic sequelae of cross-talk between the beta2-receptor and phospholipase C.