Merck
  • Bimodal strategy for excellent audiological rehabilitation in a subject with a novel nonsense mutation of the SLC26A4 gene: A case report.
Bimodal strategy for excellent audiological rehabilitation in a subject with a novel nonsense mutation of the SLC26A4 gene: A case report.
International journal of pediatric otorhinolaryngology (2020-04-07)
Rita Malesci, Roberta Russo, Carmela Monzillo, Carla Laria, Virginia Corvino, Gennaro Auletta, Achille Iolascon, Annamaria Franzè
ABSTRACT
Sensorineural hearing loss is a heterogeneous disease caused by mutations in many genes. However, in the presence of enlarged vestibular aqueduct, it is frequently associated with mutations in the solute carrier family 26 member 4 (SLC26A4), a gene causative of a syndromic form (Pendred) as well as a non-syndromic form of hearing loss (DFNB4). We describe a clinical case presenting bilateral sensorineural hearing loss and enlarged vestibular aqueduct in which a novel homozygous SLC26A4 mutation was identified. Despite a late diagnosis of hearing loss, a peculiar rehabilitation therapy strategy was identified that provided excellent results.