Merck
  • Epithelial Cells in Endometriosis and Adenomyosis Upregulate STING Expression.
Epithelial Cells in Endometriosis and Adenomyosis Upregulate STING Expression.
Reproductive sciences (Thousand Oaks, Calif.) (2020-02-13)
Hong Qu, Lihong Li, Tian-Li Wang, Tamer Seckin, James Segars, Ie-Ming Shih
ABSTRACT
In response to cytosolic DNA, stimulator of interferon gene (STING) initiates and orchestrates host's innate immunity by inducing type I interferon. Since endometriosis is a chronic inflammatory disorder, we sought to determine whether STING pathway is activated in ectopic endometrium in comparison to eutopic endometrium. Immunohistochemistry was employed in evaluating the expression levels of STING in normal endometrium, endometriosis, and adenomyosis. The density of CD45+ intraepithelial lymphocytes was correlated with STING expression levels. A total of 39 cases of endometriosis and/or adenomyosis with normal endometrium were analyzed. Among them, 32 had adenomyosis, 26 had endometriosis, and 19 have both lesions. STING protein expression is mainly evident in the cytoplasm of epithelial cells but much less in stromal cells. Based on H-score, we found that the STING expression levels were significantly higher in the epithelial cells of adenomyosis and endometriosis than in eutopic endometrium (132.7 ± 12.20, 119.6 ± 12.57 vs. 19.74 ± 5.96, p < 0.0001). There was no significant difference in STING expression level between endometriosis and adenomyosis. More intraepithelial lymphocytes were detected in endometriosis and adenomyosis lesions than endometrium (5.60 ± 0.70%, 4.95 ± 0.54% vs. 1.25 ± 0.12%, p < 0.0001). A positive correlation between STING expression and intraepithelial lymphocytic infiltrate was observed (p < 0.0001). In summary, STING was upregulated in the epithelium of ectopic endometrium as compared to eutopic endometrium. Its expression levels correlate with the degree of intraepithelial lymphocyte infiltration, suggesting a role in promoting chronic inflammation of ectopic endometrium.
Sigma-Aldrich
Anti-GAPDH, affinity isolated antibody