Merck
  • Simplifying the creation of hollow metallic nanostructures: one-pot synthesis of hollow palladium/platinum single-crystalline nanocubes.
Simplifying the creation of hollow metallic nanostructures: one-pot synthesis of hollow palladium/platinum single-crystalline nanocubes.
Angewandte Chemie (International ed. in English) (2009-05-16)
Xiaoqing Huang, Huihui Zhang, Changyou Guo, Zhiyou Zhou, Nanfeng Zheng
ABSTRACT
Efficiency simplified: A synthetic strategy has been developed to prepare single-crystalline hollow Pd/Pt nanocubes (right, see picture; left: nanocubes). Compared to the solid Pd/Pt nanocubes of similar sizes, the hollow Pd/Pt nanocubes increase accessible surface area and therefore improve electrocatalytic activity in formic acid oxidation.
Sigma-Aldrich
Palladium(II) acetylacetonate, ≥99.9% trace metals basis