Merck
  • Loss of RIG-I leads to a functional replacement with MDA5 in the Chinese tree shrew.
Loss of RIG-I leads to a functional replacement with MDA5 in the Chinese tree shrew.
Proceedings of the National Academy of Sciences of the United States of America (2016-09-14)
Ling Xu, Dandan Yu, Yu Fan, Li Peng, Yong Wu, Yong-Gang Yao
ABSTRACT
The function of the RIG-I-like receptors (RLRs; including RIG-I, MDA5, and LGP2) as key cytoplasmic sensors of viral pathogen-associated molecular patterns (PAMPs) has been subjected to numerous pathogenic challenges and has undergone a dynamic evolution. We found evolutionary evidence that RIG-I was lost in the Chinese tree shrew lineage. Along with the loss of RIG-I, both MDA5 (tMDA5) and LGP2 (tLGP2) have undergone strong positive selection in the tree shrew. tMDA5 or tMDA5/tLGP2 could sense Sendai virus (an RNA virus posed as a RIG-I agonist) for inducing type I IFN, although conventional RIG-I and MDA5 were thought to recognize distinct RNA structures and viruses. tMDA5 interacted with adaptor tMITA (STINGTMEM173/ERIS), which was reported to bind only with RIG-I. The positively selected sites in tMDA5 endowed the substitute function for the lost RIG-I. These findings provided insights into the adaptation and functional diversity of innate antiviral activity in vertebrates.
Sigma-Aldrich
Anti-MDA5 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Protease Inhibitor Cocktail Set I, A cocktail of five protease inhibitors that will inhibit a broad range of proteases and esterases. Supplied with a data sheet.