Merck
  • Explosive-driven double-blast exposure: molecular, histopathological, and behavioral consequences.
Explosive-driven double-blast exposure: molecular, histopathological, and behavioral consequences.
Scientific reports (2020-10-17)
Erin K Murphy, Diego Iacono, Hongna Pan, Jamie B Grimes, Steven Parks, Sorana Raiciulescu, Fabio Leonessa, Daniel P Perl
ABSTRACT
Traumatic brain injury generated by blast may induce long-term neurological and psychiatric sequelae. We aimed to identify molecular, histopathological, and behavioral changes in rats 2 weeks after explosive-driven double-blast exposure. Rats received two 30-psi (~ 207-kPa) blasts 24 h apart or were handled identically without blast. All rats were behaviorally assessed over 2 weeks. At Day 15, rats were euthanized, and brains removed. Brains were dissected into frontal cortex, hippocampus, cerebellum, and brainstem. Western blotting was performed to measure levels of total-Tau, phosphorylated-Tau (pTau), amyloid precursor protein (APP), GFAP, Iba1, αII-spectrin, and spectrin breakdown products (SBDP). Kinases and phosphatases, correlated with tau phosphorylation were also measured. Immunohistochemistry for pTau, APP, GFAP, and Iba1 was performed. pTau protein level was greater in the hippocampus, cerebellum, and brainstem and APP protein level was greater in cerebellum of blast vs control rats (p < 0.05). GFAP, Iba1, αII-spectrin, and SBDP remained unchanged. No immunohistochemical or neurobehavioral changes were observed. The dissociation between increased pTau and APP in different regions in the absence of neurobehavioral changes 2 weeks after double blast exposure is a relevant finding, consistent with human data showing that battlefield blasts might be associated with molecular changes before signs of neurological and psychiatric disorders manifest.
Sigma-Aldrich
Protease Inhibitor Cocktail powder, for general use, lyophilized powder
Sigma-Aldrich
Anti-PP2A Antibody, B subunit, clone 2G9, ascites fluid, clone 2G9, Upstate®
Sigma-Aldrich
Anti-APP A4 Antibody, a.a. 66-81 of APP {NT}, clone 22C11, clone 22C11, Chemicon®, from mouse
Sigma-Aldrich
Rabbit Anti-Chicken IgG Antibody, HRP conjugate, 0.8 mg/mL, Chemicon®
Sigma-Aldrich
Anti-Spectrin alpha chain (nonerythroid) Antibody, clone AA6, clone AA6, Chemicon®, from mouse
Sigma-Aldrich
Anti-GAPDH Antibody, from chicken, purified by affinity chromatography