Merck
  • Toxic effects of bisphenol A and its analogues on cyanobacteria Anabaena variabilis and Microcystis aeruginosa.
Toxic effects of bisphenol A and its analogues on cyanobacteria Anabaena variabilis and Microcystis aeruginosa.
Chemosphere (2020-12-11)
Karolina Czarny, Barbara Krawczyk, Dominik Szczukocki
ABSTRACT
In the last decades, the use of bisphenol A has attracted global attention resulting from its actions as an endocrine disrupting compound. In this regard, various bisphenol analogues have been manufactured as a replacement for this compound in consumer products. As a result of the high production volumes, different bisphenol analogues are entered into the terrestrial and aquatic environment, which consequently leads to their increasing contamination and may pose serious risk to organisms. Nevertheless, only few studies have reported on the toxic effect of bisphenol analogues on phytoplankton. Therefore, in this study, the anticyanobacterial activity of six bisphenol analogues and their mixture were investigated for the first time. Bisphenol AF, bisphenol B and bisphenol C (14 d, EC50 12.88-54.87 mg L-1) exhibit more toxic effect to both tested species in comparison to bisphenol A (14 d, EC50 55.27-78.96 mg L-1). Moreover, data show that mixture of bisphenol analogues (14 d, EC50 32.32-60.88 mg L-1) exhibit toxic effect similar to or even stronger than that of bisphenol A. The toxic effect of bisphenol analogues, singly and in combination on the growth of both cyanobacteria species was arranged in the following order: bisphenol AF > bisphenol C> bisphenol B> bisphenol A> bisphenol E> bisphenol BP and bisphenol B> bisphenol AF > bisphenol C> bisphenol A> bisphenol E> bisphenol BP for Anabaena variabilis and Microcystis aeruginosa, respectively. This research aims to assure a basic understanding of the toxic effects of bisphenol analogues on cyanobacteria and provides a more comprehensive view on environmental risk assessment.
Product Description
Supelco
Bisphenol BP, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Bisphenol A, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
4,4′-(Hexafluoroisopropylidene)diphenol, 97%
Pricing and availability is not currently available.