Merck
Dynamic Cardiolipin Synthesis Is Required for CD8+ T Cell Immunity.
Cell metabolism (2020-12-03)
Mauro Corrado, Joy Edwards-Hicks, Matteo Villa, Lea J Flachsmann, David E Sanin, Maaike Jacobs, Francesc Baixauli, Michal Stanczak, Eve Anderson, Mai Azuma, Andrea Quintana, Jonathan D Curtis, Thomas Clapes, Katarzyna M Grzes, Agnieszka M Kabat, Ryan Kyle, Annette E Patterson, Ramon Klein Geltink, Borko Amulic, Colin G Steward, Douglas Strathdee, Eirini Trompouki, David O'Sullivan, Edward J Pearce, Erika L Pearce
ABSTRACT
Mitochondria constantly adapt to the metabolic needs of a cell. This mitochondrial plasticity is critical to T cells, which modulate metabolism depending on antigen-driven signals and environment. We show here that de novo synthesis of the mitochondrial membrane-specific lipid cardiolipin maintains CD8+ T cell function. T cells deficient for the cardiolipin-synthesizing enzyme PTPMT1 had reduced cardiolipin and responded poorly to antigen because basal cardiolipin levels were required for activation. However, neither de novo cardiolipin synthesis, nor its Tafazzin-dependent remodeling, was needed for T cell activation. In contrast, PTPMT1-dependent cardiolipin synthesis was vital when mitochondrial fitness was required, most notably during memory T cell differentiation or nutrient stress. We also found CD8+ T cell defects in a small cohort of patients with Barth syndrome, where TAFAZZIN is mutated, and in a Tafazzin-deficient mouse model. Thus, the dynamic regulation of a single mitochondrial lipid is crucial for CD8+ T cell immunity.
Sigma-Aldrich
Anti-PTPMT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
Alexidine dihydrochloride, ≥95% (HPLC)
Sigma-Aldrich
Ionomycin calcium salt, Ready Made Solution, from Streptomyces conglobatus, 1 mM in DMSO
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Anti-Rat IgG (Fc specific)-Biotin antibody produced in goat, affinity isolated antibody, lyophilized powder