Merck
  • Stereochemical sensitivity of the human UDP-glucuronosyltransferases 2B7 and 2B17.
Stereochemical sensitivity of the human UDP-glucuronosyltransferases 2B7 and 2B17.
Journal of medicinal chemistry (2006-03-03)
Ingo Bichlmaier, Antti Siiskonen, Moshe Finel, Jari Yli-Kauhaluoma
ABSTRACT
A set of 28 enantiomers comprising rigid and flexible secondary alcohols was synthesized by the asymmetric Corey-Bakshi-Shibata reduction. The enantiomerically pure alcohols were subjected to enzymatic glucuronidation assays employing the human UDP-glucuronosyltransferases (UGTs) 2B7 and 2B17. Both UGTs displayed high levels of stereoselectivity, favoring the conjugation of the (R)-enantiomers over their respective (S)-stereoisomers at eudismic ratios up to 256. The spatial arrangement of the hydroxy group determined the diastereoselectivity of the UGT2B17-catalyzed reaction in agreement with Pfeiffer's rule (eudismic activity quotient = 0.83 +/- 0.14). Inhibition studies revealed that the enantiomers had similar affinities toward the enzymes. The diastereoselectivity of the UGT-catalyzed conjugation stemmed, therefore, from the arrangement of the substrates in the catalytic site, rather than from distinct affinities toward the enzymes. Taken together, this study showed that metabolic enzymes that are generally conceived to be rather "flexible" in nature are capable of displaying high levels of chiral distinction.
Sigma-Aldrich
(R)-(−)-1-Indanol, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
(S)-(−)-α−Methyl-2-naphthalenemethanol, 98%
Pricing and availability is not currently available.