Merck
  • A combined in vitro in-silico approach to predict the oral bioavailability of borderline BCS Class II/IV weak base albendazole and its main metabolite albendazole sulfoxide.
A combined in vitro in-silico approach to predict the oral bioavailability of borderline BCS Class II/IV weak base albendazole and its main metabolite albendazole sulfoxide.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2020-09-17)
Maximo Pettarin, Michael B Bolger, Maja Chronowska, Edmund S Kostewicz
ABSTRACT
The aim of this study was to use a combined in vitro-in silico approach to develop a physiologically based pharmacokinetic model (PBPK) that predicts the bioavailability of albendazole (ABZ), a BCS class II/IV lipophilic weak base, and simulates its main metabolite albendazole sulphoxide (ABZSO) after oral administration of the current marketed dose of 400 mg in the fasted state. In vitro data was collected from solubility and dissolution tests performed with biorelevant media and transfer tests were carried out to evaluate the supersaturation and precipitation characteristics of ABZ upon gastric emptying. These in vitro results were used as biopharmaceutical inputs together with ABZ physicochemical properties including also permeability and in vitro metabolism data and information gathered from different clinical trials reported in the literature, were used to enable PBPK models to be developed using GastroPlus™ (version 9.7). As expected for this weak base with pKa = 3.6, ABZ exhibited a pronounced pH dependent solubility, with the solubility and extent of dissolution being greater at gastric pH and dropping significantly in the intestinal environment suggesting supersaturation and precipitation upon gastric emptying, which was confirmed by the transfer model experiments. PBPK models were set up for heathy volunteers using a full PBPK modeling approach and by implementing dynamic fluid volumes in the ACAT gut physiology in GastroPlus™. When coupling in vitro data (solubility values, dissolution rate and precipitation rate constant, etc.) for ABZ and with fitted values for the Vdss and liver systemic clearance of the sulfoxide metabolite to the PBPK model, the simulated profiles successfully predicated plasma concentrations of ABZ at 400 mg dose and simulated ABZSO at different ABZ dose levels and with different study populations, indicating the usefulness of combing in vitro biorelevant tools with PBPK modeling for the accurate prediction of ABZ bioavailability. The results obtained in this study also helped confirm that ABZ behaves as a BCS class IV compound.
Sigma-Aldrich
Albendazole sulfoxide, ≥97% (HPLC)