Merck
  • A sandwich-type cluster containing Ge@Pd3 planar fragment flanked by aromatic nonagermanide caps.
A sandwich-type cluster containing Ge@Pd3 planar fragment flanked by aromatic nonagermanide caps.
Nature communications (2020-10-22)
Hong-Lei Xu, Nikolay V Tkachenko, Zi-Chuan Wang, Wei-Xing Chen, Lei Qiao, Alvaro Muñoz-Castro, Alexander I Boldyrev, Zhong-Ming Sun
ABSTRACT
Sandwich-type clusters with the planar fragment containing a heterometallic sheet have remained elusive. In this work, we introduce the [K(2,2,2-crypt)]4{(Ge9)2[η6-Ge(PdPPh3)3]} complex that contains a heterometallic sandwich fragment. The title compound is structurally characterized by means of single-crystal X-ray diffraction, which reveals the presence of an unusual heteroatomic metal planar fragment Ge@Pd3. The planar fragment contains a rare formal zerovalent germanium core and a peculiar bonding mode of sp2-Ge@(PdPPh3)3 trigonal planar structure, whereas the nonagermanide fragments act as capping ligands. The chemical bonding pattern of the planar fragment consists of three 2c-2e Pd-Ge σ-bonds attaching Pd atoms to the core Ge atom, while the binding between the planar fragment and the aromatic Ge9 ligands is provided by six 2c-2e Pd-Ge σ-bonds and two delocalized 4c-2e σ-bonds. The synthesized cluster represents a rare example of a sandwich compound with the heteroatomic metal planar fragment and inorganic aromatic capping ligands.
Sigma-Aldrich
Tetrakis(triphenylphosphine)palladium(0), 99%
Pricing and availability is not currently available.