Merck
  • The Natural Compound Notopterol Binds and Targets JAK2/3 to Ameliorate Inflammation and Arthritis.
The Natural Compound Notopterol Binds and Targets JAK2/3 to Ameliorate Inflammation and Arthritis.
Cell reports (2020-09-17)
Qiong Wang, Xin Zhou, Long Yang, Yongjian Zhao, Zhihuan Chew, Jun Xiao, Chang Liu, Xin Zheng, Yuxiao Zheng, Qi Shi, Qianqian Liang, Yongjun Wang, Hongyan Wang
ABSTRACT
The traditional Chinese medicinal herb Notopterygium incisum Ting ex H.T. Chang has anti-rheumatism activity, and a mass spectrometry assay of patients' serum after administration of the herb revealed that notopterol is the most abundant component enriched. However, the functions of notopterol and its molecular target in rheumatoid arthritis (RA) treatment remain unknown. Here, we show in different RA mouse strains that both oral and intraperitoneal administration of notopterol result in significant therapeutic effects. Mechanistically, notopterol directly binds Janus kinase (JAK)2 and JAK3 kinase domains to inhibit JAK/signal transducers and activators of transcription (JAK-STAT) activation, leading to reduced production of inflammatory cytokines and chemokines. Critically, combination therapy using both notopterol and tumor necrosis factor (TNF) blocker results in enhanced therapeutic effects compared to using TNF blocker alone. We demonstrate that notopterol ameliorates RA pathology by targeting JAK-STAT signaling, raising the possibility that notopterol could be effective in treating other diseases characterized by aberrant JAK-STAT signaling pathway.
Product Description
Notopterol, phyproof® Reference Substance
Millipore
Brewer thioglycollate medium, suitable for microbiology, NutriSelect® Plus