Merck
  • Polyunsaturated Fatty Acid Desaturation Is a Mechanism for Glycolytic NAD+ Recycling.
Polyunsaturated Fatty Acid Desaturation Is a Mechanism for Glycolytic NAD+ Recycling.
Cell metabolism (2019-01-29)
Wondong Kim, Amy Deik, Clicerio Gonzalez, Maria Elena Gonzalez, Feifei Fu, Michele Ferrari, Claire L Churchhouse, Jose C Florez, Suzanne B R Jacobs, Clary B Clish, Eugene P Rhee
ABSTRACT
The reactions catalyzed by the delta-5 and delta-6 desaturases (D5D/D6D), key enzymes responsible for highly unsaturated fatty acid (HUFA) synthesis, regenerate NAD+ from NADH. Here, we show that D5D/D6D provide a mechanism for glycolytic NAD+ recycling that permits ongoing glycolysis and cell viability when the cytosolic NAD+/NADH ratio is reduced, analogous to lactate fermentation. Although lesser in magnitude than lactate production, this desaturase-mediated NAD+ recycling is acutely adaptive when aerobic respiration is impaired in vivo. Notably, inhibition of either HUFA synthesis or lactate fermentation increases the other, underscoring their interdependence. Consistent with this, a type 2 diabetes risk haplotype in SLC16A11 that reduces pyruvate transport (thus limiting lactate production) increases D5D/D6D activity in vitro and in humans, demonstrating a chronic effect of desaturase-mediated NAD+ recycling. These findings highlight key biologic roles for D5D/D6D activity independent of their HUFA end products and expand the current paradigm of glycolytic NAD+ regeneration.
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
NHI-2, ≥98% (HPLC)
Sigma-Aldrich
SC-26196, ≥98% (HPLC)