Merck
PARP inhibition: PARP1 and beyond.
Nature reviews. Cancer (2010-03-05)
Michèle Rouleau, Anand Patel, Michael J Hendzel, Scott H Kaufmann, Guy G Poirier
ABSTRACT
Recent findings have thrust poly(ADP-ribose) polymerases (PARPs) into the limelight as potential chemotherapeutic targets. To provide a framework for understanding these recent observations, we review what is known about the structures and functions of the family of PARP enzymes, and then outline a series of questions that should be addressed to guide the rational development of PARP inhibitors as anticancer agents.