Merck
  • Calcium-sensing receptor deletion in the mouse esophagus alters barrier function.
Calcium-sensing receptor deletion in the mouse esophagus alters barrier function.
American journal of physiology. Gastrointestinal and liver physiology (2019-11-12)
Nazih L Nakhoul, Chia-Ling Tu, Karen L Brown, M Toriqul Islam, Anna G Hodges, Solange M Abdulnour-Nakhoul
ABSTRACT
Calcium-sensing receptor (CaSR) is the molecular sensor by which cells respond to small changes in extracellular Ca2+ concentrations. CaSR has been reported to play a role in glandular and fluid secretion in the gastrointestinal tract and to regulate differentiation and proliferation of skin keratinocytes. CaSR is present in the esophageal epithelium, but its role in this tissue has not been defined. We deleted CaSR in the mouse esophagus by generating keratin 5 CreER;CaSRFlox+/+compound mutants, in which loxP sites flank exon 7 of CaSR gene. Recombination was initiated with multiple tamoxifen injections, and we demonstrated exon 7 deletion by PCR analysis of genomic DNA. Quantitative real-time PCR and Western blot analyses showed a significant reduction in CaSR mRNA and protein expression in the knockout mice (EsoCaSR-/-) as compared with control mice. Microscopic examination of EsoCaSR-/- esophageal tissues showed morphological changes including elongation of the rete pegs, abnormal keratinization and stratification, and bacterial buildup on the luminal epithelial surface. Western analysis revealed a significant reduction in levels of adherens junction proteins E-cadherin and β catenin and tight junction protein claudin-1, 4, and 5. Levels of small GTPase proteins Rac/Cdc42, involved in actin remodeling, were also reduced. Ussing chamber experiments showed a significantly lower transepithelial resistance in knockout (KO) tissues. In addition, luminal-to-serosal-fluorescein dextran (4 kDa) flux was higher in KO tissues. Our data indicate that CaSR plays a role in regulating keratinization and cell-cell junctional complexes and is therefore important for the maintenance of the barrier function of the esophagus.NEW & NOTEWORTHY The esophageal stratified squamous epithelium maintains its integrity by continuous proliferation and differentiation of the basal cells. Here, we demonstrate that deletion of the calcium-sensing receptor, a G protein-coupled receptor, from the basal cells disrupts the structure and barrier properties of the epithelium.
Sigma-Aldrich
Anti-Calcium Sensing Receptor antibody, Mouse monoclonal, clone HL1499 (6D4-3F4-6C4), purified from hybridoma cell culture
Sigma-Aldrich
Anti-Ca++ Sensing Receptor Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Hematoxylin Powder (C.I. 75290), HARLECO® Nuclear staining