Merck
  • Retinoic acid promotes metabolic maturation of human Embryonic Stem Cell-derived Cardiomyocytes.
Retinoic acid promotes metabolic maturation of human Embryonic Stem Cell-derived Cardiomyocytes.
Theranostics (2020-08-31)
Shumei Miao, Dandan Zhao, Xiaoxiao Wang, Xuan Ni, Xing Fang, Miao Yu, Lingqun Ye, Jingsi Yang, Hongchun Wu, Xinglong Han, Lina Qu, Lei Li, Feng Lan, Zhenya Shen, Wei Lei, Zhen-Ao Zhao, Shijun Hu
ABSTRACT
Cardiomyocytes differentiated from human embryonic stem cells (hESCs) represent a promising cell source for heart repair, disease modeling and drug testing. However, improving the differentiation efficiency and maturation of hESC-derived cardiomyocytes (hESC-CMs) is still a major concern. Retinoic acid (RA) signaling plays multiple roles in heart development. However, the effects of RA on cardiomyocyte differentiation efficiency and maturation are still unknown. Methods: RA was added at different time intervals to identify the best treatment windows for cardiomyocyte differentiation and maturation. The efficiency of cardiomyocyte differentiation was detected by quantitative real-time PCR and flow cytometry. Cardiomyocytes maturation was detected by immunofluorescence staining, metabolic assays and patch clamp to verify structural, metabolic and electrophysiological maturation, respectively. RNA sequencing was used for splicing analysis. Results: We found that RA treatment at the lateral mesoderm stage (days 2-4) significantly improved cardiomyocyte differentiation, as evidenced by the upregulation of TNNT2, NKX2.5 and MYH6 on day 10 of differentiation. In addition, flow cytometry showed that the proportion of differentiated cardiomyocytes in the RA-treated group was significantly higher than that in control group. RA treatment on days 15-20 increased cardiomyocyte area, sarcomere length, multinucleation and mitochondrial copy number. RNA sequencing revealed RA promoted RNA isoform switch to the maturation-related form. Meanwhile, RA promoted electrophysiological maturation and calcium handling of hESC-CMs. Importantly, RA-treated cardiomyocytes showed decreased glycolysis and enhanced mitochondrial oxidative phosphorylation, with the increased utilization of fatty acid and exogenous pyruvate but not glutamine. Conclusion: Our data indicated that RA treatment at an early time window (days 2-4) promotes the efficiency of cardiomyocyte differentiation and that RA treatment post beating (days 15-20) promotes cardiomyocyte maturation. The biphasic effects of RA provide new insights for improving cardiomyocyte differentiation and quality.
Sigma-Aldrich
bisBenzimide H 33342 trihydrochloride, for fluorescence, ≥97.0% (HPLC)
Sigma-Aldrich
Sodium DL-lactate solution, syrup, BioReagent, 60 % (w/w), synthetic, suitable for cell culture
Sigma-Aldrich
(+)-Etomoxir sodium salt hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
L-Carnitine hydrochloride, synthetic, ≥98%
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Sodium palmitate, ≥98.5%
Sigma-Aldrich
IWR-1, ≥98% (HPLC)
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder