Merck
  • Rapid detection of microbiota cell type diversity using machine-learned classification of flow cytometry data.
Rapid detection of microbiota cell type diversity using machine-learned classification of flow cytometry data.
Communications biology (2020-07-17)
Birge D Özel Duygan, Noushin Hadadi, Ambrin Farizah Babu, Markus Seyfried, Jan R van der Meer
ABSTRACT
The study of complex microbial communities typically entails high-throughput sequencing and downstream bioinformatics analyses. Here we expand and accelerate microbiota analysis by enabling cell type diversity quantification from multidimensional flow cytometry data using a supervised machine learning algorithm of standard cell type recognition (CellCognize). As a proof-of-concept, we trained neural networks with 32 microbial cell and bead standards. The resulting classifiers were extensively validated in silico on known microbiota, showing on average 80% prediction accuracy. Furthermore, the classifiers could detect shifts in microbial communities of unknown composition upon chemical amendment, comparable to results from 16S-rRNA-amplicon analysis. CellCognize was also able to quantify population growth and estimate total community biomass productivity, providing estimates similar to those from 14C-substrate incorporation. CellCognize complements current sequencing-based methods by enabling rapid routine cell diversity analysis. The pipeline is suitable to optimize cell recognition for recurring microbiota types, such as in human health or engineered systems.
Sigma-Aldrich
Bis(diphenylphosphino)methane, 97%