Merck
  • A Global Study by 1H NMR Spectroscopy and SPME-GC/MS of the in Vitro Digestion of Virgin Flaxseed Oil Enriched or not with Mono-, Di- or Tri-Phenolic Derivatives. Antioxidant Efficiency of These Compounds.
A Global Study by 1H NMR Spectroscopy and SPME-GC/MS of the in Vitro Digestion of Virgin Flaxseed Oil Enriched or not with Mono-, Di- or Tri-Phenolic Derivatives. Antioxidant Efficiency of These Compounds.
Antioxidants (Basel, Switzerland) (2020-04-25)
Jon Alberdi-Cedeño, María L Ibargoitia, María D Guillén
ABSTRACT
The effect of enriching virgin flaxseed oil with dodecyl gallate, hydroxytyrosol acetate or gamma-tocopherol on its in vitro digestion is studied by means of proton nuclear magnetic resonance and solid phase microextraction followed by gas chromatography/mass spectrometry. The extent and pattern of the lipolysis reached in each sample is analyzed, as is the bioaccessibility of the main oil components. None of the phenolic compounds provokes inhibition of the lipase activity and all of them reduce the lipid oxidation degree caused by the in vitro digestion and the bioaccessibility of oxidation compounds. The antioxidant efficiency of the three tested phenols is in line with the number of phenolic groups in its molecule, and is dose-dependent. The concentration of some minor oil components such as terpenes, sesquiterpenes, cycloartenol and 24-methylenecycloartenol is not modified by in vitro digestion. Contrarily, gamma-tocopherol shows very low in vitro bioaccessibility, probably due to its antioxidant behavior, although this increases with enrichment of the phenolic compounds. Oxidation is produced during in vitro digestion even in the presence of a high concentration of gamma-tocopherol, which remains bioaccessible after digestion in the enriched samples of this compound.
Sigma-Aldrich
Suplatast tosylate, ≥98% (HPLC)
Sigma-Aldrich
Cycloartenol, ≥90% (GC)
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥400 units/mg protein
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Microvascular Endothelial Cell Basal Medium (500ml)
Sigma-Aldrich
3,3′-Diiodo-L-thyronine (T2) hydrochloride, 98% (CP)
Millipore
Bile bovine, dried, unfractionated
Sigma-Aldrich
Pancreatin from porcine pancreas, 4 × USP specifications