Merck
  • Pesticides in aquatic environments and their removal by adsorption methods.
Pesticides in aquatic environments and their removal by adsorption methods.
Chemosphere (2020-04-11)
Amin Mojiri, John L Zhou, Brett Robinson, Akiyoshi Ohashi, Noriatsu Ozaki, Tomonori Kindaichi, Hossein Farraji, Mohammadtaghi Vakili
ABSTRACT
Although pesticides are widely used in agriculture, industry and households, they pose a risk to human health and ecosystems. Based on target organisms, the main types of pesticides are herbicides, insecticides and fungicides, of which herbicides accounted for 46% of the total pesticide usage worldwide. The movement of pesticides into water bodies occurs through run-off, spray drift, leaching, and sub-surface drainage, all of which have negative impacts on aquatic environments and humans. We sought to define the critical factors affecting the fluxes of contaminants into receiving waters. We also aimed to specify the feasibility of using sorbents to remove pesticides from waterways. In Karun River in Iran (1.21 × 105 ng/L), pesticide concentrations are above regulatory limits. The concentration of pesticides in fish can reach 26.1 × 103 μg/kg, specifically methoxychlor herbicide in Perca fluviatilis in Lithuania. During the last years, research has focused on elimination of organic pollutants, such as pesticides, from aqueous solution. Pesticide adsorption onto low-cost materials can effectively remediate contaminated waters. In particular, nanoparticle adsorbents and carbon-based adsorbents exhibit high performance (nearly 100%) in removing pesticides from water bodies.
Product Description
Supelco
Metazachlor, PESTANAL®, analytical standard
Supelco
Acetochlor, PESTANAL®, analytical standard
Supelco
Fenobucarb, PESTANAL®, analytical standard
Supelco
Mecoprop-P, PESTANAL®, analytical standard
Supelco
Propanil, PESTANAL®, analytical standard
Supelco
Isoprothiolane, PESTANAL®, analytical standard
Supelco
Esfenvalerate, PESTANAL®, analytical standard
Supelco
Molinate, PESTANAL®, analytical standard
Supelco
Dichlorprop, PESTANAL®, analytical standard
Supelco
Pirimicarb, PESTANAL®, analytical standard
Sigma-Aldrich
1H-1,2,3-Triazole, 97%
Supelco
Oxadiazon, PESTANAL®, analytical standard