Merck
  • Adenosine kinase inhibition attenuates ischemia reperfusion-induced acute kidney injury.
Adenosine kinase inhibition attenuates ischemia reperfusion-induced acute kidney injury.
Life sciences (2020-06-17)
Wei Cao, Huiting Wan, Lin Wu, Xiaofei An, Qing Li, Jiaxing Dong, Suyan Duan, Zhimin Huang, Bo Zhang, Changying Xing, Yanggang Yuan
ABSTRACT
Acute kidney injury (AKI) has a high morbidity and mortality, and there is no targeted treatment yet. One of the main causes of AKI is ischemia-reperfusion (IR). Increased release of adenosine under stress and hypoxia exerts anti-inflammatory and antioxidant effects. Adenosine kinase (ADK) is an important enzyme that eliminates adenosine in cells, and can maintain low adenosine concentration in cells. Our previous studies have shown that pretreatment of adenosine kinase inhibitor ABT-702 could markedly attenuate cisplatin-induced nephrotoxicity both in vivo and in vitro. This study is designed to investigate the effect of ADK inhibition on IR-induced AKI. The results showed that ADK expression was positively correlated with the degree of renal tubular injury, which suggested that the degree of ADK inhibition reflected the severity of acute tubular necrosis. In vivo, ADK inhibitor could reduce IR-induced renal injury, which might play a protective role by increasing tissue adenosine level, inhibiting oxidative stress, and reducing cell apoptosis. In HK2 cells, cobaltous dichloride (CoCl2) increased the level of oxidative stress, up-regulated the production of pro-inflammatory factor, and induced apoptosis, ADK inhibition could alleviate the above damaging effects. Moreover, the anti-apoptotic effect exerted by ADK inhibition was independent of inosine. In summary, our results support the idea that ADK inhibition has protective effects on IR-induced AKI. Adenosine kinase inhibition might provide a new target for AKI prevention and treatment.
Sigma-Aldrich
MISSION® esiRNA, targeting human ADK
Sigma-Aldrich
Inosine, ≥99% (HPLC)