- Antibiotic resistance markers for genetic manipulations of Leptospira spp.
Antibiotic resistance markers for genetic manipulations of Leptospira spp.
Applied and environmental microbiology (2010-06-01)
Dante Poggi, Priscila Oliveira de Giuseppe, Mathieu Picardeau
PMID20511419
ABSTRACT
We measured the frequency of appearance of spontaneous mutants resistant to gentamicin, kanamycin, streptomycin, and spectinomycin in saprophytic and pathogenic Leptospira strains. The mutations responsible for the spontaneous resistance to streptomycin and spectinomycin were identified in the rpsL and rrs genes, respectively. We also generated a gentamicin resistance cassette that allows the use of a third selectable marker in leptospires. These results may facilitate further advances in gene transfer systems in Leptospira spp.
Sigma-Aldrich
Spectinomycin ready made solution, 100 mg/mL in DMSO/H2O, 1:1, Cell culture tested