Merck
Role of PA2G4P4 pseudogene in bladder cancer tumorigenesis.
Biology (2020-04-05)
Laura Pisapia, Sara Terreri, Pasquale Barba, Marianna Mastroianni, Maria Donnini, Vincenzo Mercadante, Alessandro Palmieri, Paolo Verze, Vincenzo Mirone, Vincenzo Altieri, Gianluigi Califano, Giovanna Lucia Liguori, Maria Strazzullo, Amelia Cimmino, Giovanna Del Pozzo
ABSTRACT
Many pseudogenes possess biological activities and play important roles in the pathogenesis of various types of cancer including bladder cancer (BlCa), which still lacks suitable molecular biomarkers. Recently, pseudogenes were found to be significantly enriched in a pan-cancer classification based on the Cancer Genome Atlas gene expression data. Among them, the top-ranking pseudogene was the proliferation-associated 2G4 pseudogene 4 (PA2G4P4). Genomic and transcript features of PA2G4P4 were determined by GeneBank database analysis followed by 5' RACE experiments. Therefore, we conducted a retrospective molecular study on a cohort of 45 patients of BlCa. PA2G4P4 expression was measured by RT-qPCR, whereas PA2G4P4 transcript distribution was analyzed by in situ hybridization on both normal and cancerous histological sections and compared to the immunolocalization of its parental PA2G4/EBP1 protein. Finally, we tested the effects of PA2G4P4 depletion on proliferation, migration, and death of BlCa cells. We showed for the first time PA2G4P4 overexpression in BlCa tissues and in cell lines. PA2G4P4 distribution strictly overlaps PA2G4/EBP1 protein localization. Moreover, we showed that PA2G4P4 knockdown affects both proliferation and migration of BlCa cells, highlighting its potential oncogenic role. PA2G4P4 may play a functional role as an oncogene in BlCa development, suggesting it as a good candidate for future investigation and new clinical applications.
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, F(ab′)2, HRP conjugate, Chemicon®, from goat
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture