Merck
  • Development and pharmacological evaluation of ropivacaine-2-hydroxypropyl-beta-cyclodextrin inclusion complex.
Development and pharmacological evaluation of ropivacaine-2-hydroxypropyl-beta-cyclodextrin inclusion complex.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2007-11-27)
Daniele R de Araujo, Simone S Tsuneda, Cíntia M S Cereda, Fernanda Del G F Carvalho, Paulo S C Preté, Sergio A Fernandes, Fabiano Yokaichiya, Margareth K K D Franco, Irineu Mazzaro, Leonardo F Fraceto, Angélica de F A Braga, Eneida de Paula
ABSTRACT
Ropivacaine (RVC) is an enantiomerically pure local anesthetic (LA) largely used in surgical procedures, which presents physico-chemical and therapeutic properties similar to those of bupivacaine (BPV), but associated to less systemic toxicity. This study focuses on the development and pharmacological evaluation of a RVC in 2-hydroxypropyl-beta-cyclodextrin (HP-beta-CD) inclusion complex. Phase-solubility diagrams allowed the determination of the association constant between RVC and HP-beta-CD (9.46 M(-1)) and showed an increase on RVC solubility upon complexation. Release kinetics revealed a decrease on RVC release rate and reduced hemolytic effects after complexation (onset at 3.7 mM and 11.2mM for RVC and RVC HP-beta-CD, respectively) were observed. Differential scanning calorimetry (DSC), scanning electron microscopy (SEM) and X-ray analysis (X-ray) showed the formation and the morphology of the complex. Nuclear magnetic resonance (NMR) and job-plot experiments afforded data regarding inclusion complex stoichiometry (1:1) and topology. Sciatic nerve blockade studies showed that RVC HP-beta-CD was able to reduce the latency without increasing the duration of motor blockade, but prolonging the duration and intensity of the sensory blockade (p<0.001) induced by the LA in mice. These results identify the RVC HP-beta-CD complex as an effective novel approach to enhance the pharmacological effects of RVC, presenting it as a promising new anesthetic formulation.
Sigma-Aldrich
(2-Hydroxypropyl)-β-cyclodextrin, average Mw ~1,460