Merck
  • Utilization of CRISPR Interference To Validate MmpL3 as a Drug Target in Mycobacterium tuberculosis.
Utilization of CRISPR Interference To Validate MmpL3 as a Drug Target in Mycobacterium tuberculosis.
Antimicrobial agents and chemotherapy (2019-06-05)
Matthew B McNeil, Gregory M Cook
ABSTRACT
There is an urgent need for novel therapeutics to treat Mycobacterium tuberculosis infections. Genetic strategies for validating novel targets are available, yet their time-consuming nature limits their utility. Here, using MmpL3 as a model target, we report on the application of mycobacterial CRISPR interference for the rapid validation of target essentiality and compound mode of action. This strategy has the potential to rapidly accelerate tuberculosis drug discovery.
Sigma-Aldrich
SQ109, ≥98% (HPLC)