Merck
  • ER stress in antigen-presenting cells promotes NKT cell activation through endogenous neutral lipids.
ER stress in antigen-presenting cells promotes NKT cell activation through endogenous neutral lipids.
EMBO reports (2020-05-05)
Srinath Govindarajan, Eveline Verheugen, Koen Venken, Djoere Gaublomme, Margaux Maelegheer, Eva Cloots, Fien Gysens, Bruno G De Geest, Tan-Yun Cheng, D Branch Moody, Sophie Janssens, Michael Drennan, Dirk Elewaut
ABSTRACT
CD1d-restricted invariant natural killer T (iNKT) cells constitute a common glycolipid-reactive innate-like T-cell subset with a broad impact on innate and adaptive immunity. While several microbial glycolipids are known to activate iNKT cells, the cellular mechanisms leading to endogenous CD1d-dependent glycolipid responses remain largely unclear. Here, we show that endoplasmic reticulum (ER) stress in APCs is a potent inducer of CD1d-dependent iNKT cell autoreactivity. This pathway relies on the presence of two transducers of the unfolded protein response: inositol-requiring enzyme-1a (IRE1α) and protein kinase R-like ER kinase (PERK). Surprisingly, the neutral but not the polar lipids generated within APCs undergoing ER stress are capable of activating iNKT cells. These data reveal that ER stress is an important mechanism to elicit endogenous CD1d-restricted iNKT cell responses through induction of distinct classes of neutral lipids.
Sigma-Aldrich
Linoleic acid, ≥98%
Sigma-Aldrich
1,2-Dioleoyl-sn-glycerol, ≥97%
Avanti
C24:1 Galactosyl(β) Ceramide (d18:1/24:1(15Z)), Avanti Research - A Croda Brand 860546P, powder
Avanti
24:1 SM, Avanti Research - A Croda Brand, powder
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Avanti
C18 Ceramide (d18:1/18:0), Avanti Research - A Croda Brand