Merck
  • OVOL2-Mediated ZEB1 Downregulation May Prevent Promotion of Actinic Keratosis to Cutaneous Squamous Cell Carcinoma.
OVOL2-Mediated ZEB1 Downregulation May Prevent Promotion of Actinic Keratosis to Cutaneous Squamous Cell Carcinoma.
Journal of clinical medicine (2020-02-29)
Maho Murata, Takamichi Ito, Yuka Tanaka, Kazuhiko Yamamura, Kazuhisa Furue, Masutaka Furue
ABSTRACT
Progression of actinic keratosis (AK) to cutaneous squamous cell carcinoma (cSCC) is rare. Most cases of AK remain as intraepidermal lesions, owing to the suppression of the epithelial-to-mesenchymal transition (EMT). Ovo-like transcriptional repressor 1 (OVOL1) and ovo-like zinc finger 2 (OVOL2) are important modulators of EMT in some tumors, but their roles in skin tumors remain elusive. This study elucidated the roles of OVOL1/2 in AK and cSCC using 30 AK/30 cSCC clinical samples, and an A431 human SCC cell line using immunohistochemistry and molecular biological approaches. Immunohistochemically, OVOL1/2 were upregulated in AK and downregulated in cSCC. Meanwhile, EMT-related factors, vimentin and zinc finger E-box binding homeobox 1 (ZEB1) were downregulated in AK and upregulated in cSCC. Moreover, ZEB1 expression was higher in tumors in which OVOL2 expression was low. Thus, we observed an inverse association between OVOL2 and ZEB1 expression in AK and cSCC. Although knockdown of OVOL1 or OVOL2 increased the mRNA and protein levels of ZEB1, only OVOL2 knockdown increased the invasive ability of A431. In conclusion, OVOL2 inhibits ZEB1 expression and may inhibit the promotion of AK into cSCC. OVOL2/ZEB1 axis may be a potential target for preventing the development of cSCC.
Sigma-Aldrich
MISSION® esiRNA, targeting human OVOL1
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, sodium pyruvate, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture