Merck
  • Structure of H3K36-methylated nucleosome-PWWP complex reveals multivalent cross-gyre binding.
Structure of H3K36-methylated nucleosome-PWWP complex reveals multivalent cross-gyre binding.
Nature structural & molecular biology (2019-12-11)
Haibo Wang, Lucas Farnung, Christian Dienemann, Patrick Cramer
ABSTRACT
Recognition of histone-modified nucleosomes by specific reader domains underlies the regulation of chromatin-associated processes. Whereas structural studies revealed how reader domains bind modified histone peptides, it is unclear how reader domains interact with modified nucleosomes. Here, we report the cryo-electron microscopy structure of the PWWP reader domain of human transcriptional coactivator LEDGF in complex with an H3K36-methylated nucleosome at 3.2-Å resolution. The structure reveals multivalent binding of the reader domain to the methylated histone tail and to both gyres of nucleosomal DNA, explaining the known cooperative interactions. The observed cross-gyre binding may contribute to nucleosome integrity during transcription. The structure also explains how human PWWP domain-containing proteins are recruited to H3K36-methylated regions of the genome for transcription, histone acetylation and methylation, and for DNA methylation and repair.
Sigma-Aldrich
(2-Bromoethyl)trimethylammonium bromide, 98%
Pricing and availability is not currently available.