Merck
  • Regenerative effects of glycyrrhizin and/or platelet rich plasma on type-II collagen induced arthritis: Targeting autophay machinery markers, inflammation and oxidative stress.
Regenerative effects of glycyrrhizin and/or platelet rich plasma on type-II collagen induced arthritis: Targeting autophay machinery markers, inflammation and oxidative stress.
Archives of biochemistry and biophysics (2019-09-03)
Noha M Shafik, Rasha Osama El-Esawy, Darin A Mohamed, Ehsan A Deghidy, Omnia Safwat El-Deeb
ABSTRACT
Rheumatoid arthritis (RA) is a systemic chronic autoimmune disease manifested by joint destruction and deformity, hence decreasing patient's life quality. The aim of the present work is to explore the mechanistic effects of glycyrrhizin (GL)and/or platelet rich plasma (PRP) treatment on collagen induced arthritis. 75 female Wistar rats were allocated into five equal groups. Group I: control group. Group II: arthritis group (A group); arthritis was induced by type-II collagen Group III: Glycyrrhizin treated group(A + GL group), Group IV: platelet rich plasma treated group(A + PRP group)and Group V: combined treatment group(A + GL + PRP group). Hind paw joint tissue levels of high-mobility group box 1 protein (HMGB-1), beclin-1 and nuclear factor (erythroid-2)-related factor 2 (Nrf2) DNA binding activity were detected by ELISA. Activities of myeloperoxidase (MPO) and catalase enzymes were determined spectrophotometrically. mRNA expression levels of microtubule associated protein light chain 3 (LC3) was detected by quantitative real time PCR. After 8 weeks treatment, there was improvement of inflammation and autophagy biomarkers by the significant reduction of HMGB-1 and beclin-1 levels, down regulation ofLC3mRNA expression. On the other hand, we monitored restoration of the anti-oxidant status through the inhibited MPO activity besides induction of both catalase and Nrf2-DNA binding activities. It could be concluded that, the mutual use of both PRP and GL had a greater effect than each alone against arthritis which is considered a novel finding that can highlight the regenerative and ameliorative effects of this combined treatmentthus launching promising avenues for RA treatment.
Sigma-Aldrich
Glycyrrhizin
Sigma-Aldrich
Freund′s Adjuvant, Incomplete, liquid