Merck
  • Ceramide-induced impairment of endothelial function is prevented by CuZn superoxide dismutase overexpression.
Ceramide-induced impairment of endothelial function is prevented by CuZn superoxide dismutase overexpression.
Arteriosclerosis, thrombosis, and vascular biology (2004-11-06)
Sean P Didion, Frank M Faraci
ABSTRACT
Ceramide is an important intracellular second messenger that may also increase superoxide. The goal of this study was to determine whether overexpression of CuZn superoxide dismutase (SOD) protects against ceramide-induced increases in vascular superoxide and endothelial dysfunction. Carotid arteries from CuZnSOD-transgenic (CuZnSOD-Tg) and nontransgenic littermates were examined in vitro. Immunohistochemistry confirmed that CuZnSOD protein was greater in carotid artery from CuZnSOD-Tg compared with nontransgenic mice. Ceramide (N-acetyl-d-sphingosine; 1 and 10 micromol/L) produced concentration-dependent impairment (P<0.05) of vasorelaxation in response to the endothelium-dependent agonist acetylcholine (ACh) in nontransgenic mice. For example, 100 micromol/L ACh relaxed arteries from nontransgenic mice by 96+/-4% and 52+/-5% in the presence of vehicle and 10 micromol/L ceramide, respectively. In contrast, ceramide (1 or 10 micromol/L) had no effect (P>0.05) on responses of carotid artery to ACh in CuZnSOD-Tg mice. Ceramide had no effect on nitroprusside- or papaverine-induced relaxation in CuZnSOD-Tg or nontransgenic mice. Ceramide increased superoxide in arteries from nontransgenic vessels, and this effect was prevented by polyethyleneglycol-SOD (50 U/mL) or overexpression of CuZnSOD. These results suggest that ceramide-induced increases in superoxide impair endothelium-dependent relaxation, and that select overexpression of the CuZn isoform of SOD prevents ceramide-induced oxidative stress in vessels.
Sigma-Aldrich
Nω-Nitro-L-arginine, ≥98% (TLC)
Sigma-Aldrich
N-Acetyl-D-sphingosine, ≥97% (TLC), powder