Merck
  • The opposing contribution of neurotrophin-3 and nerve growth factor to orofacial heat hyperalgesia in rats.
The opposing contribution of neurotrophin-3 and nerve growth factor to orofacial heat hyperalgesia in rats.
Behavioural pharmacology (2019-10-03)
Renata Cristiane Dos Reis, Caroline Machado Kopruszinski, Carina Fernanda Mattedi Nones, Débora Assunção Aguiar, Juliana Geremias Chichorro
ABSTRACT
It has been proposed that neurotrophin-3 acts in a manner that is opposed to nerve growth factor, especially in the modulation of heat hyperalgesia. Injury to the constriction of the infraorbital nerve (CION) is a well-established model of trigeminal neuropathic pain that leads to robust heat, cold, and mechanical hyperalgesia. Here, we assessed the effect of local neurotrophin-3 treatment on CION-induced hyperalgesia, and we examined some mechanisms related to the effect of neurotrophin-3. Neurotrophin-3 (1 µg/50 µl) injected into the upper lip of CION rats caused a significant and long-lasting reduction of CION-induced heat hyperalgesia, but failed to affect cold and mechanical hyperalgesia. Increased levels of neurotrophin-3 were detected in the injured nerve at the time point that represents the peak of heat hyperalgesia. The anti-hyperalgesic effect of neurotrophin-3 was markedly reduced in the presence of an antagonist of TrkA receptors (K-252a, 1 μg/50 μl). Moreover, association of lower doses of neurotrophin-3 with an antibody anti-nerve growth factor resulted in a synergistic anti-hyperalgesic effect in CION rats. Local injection of nerve growth factor (3 µg/50 µl) or the TRPV1 agonist capsaicin (1 μg/50 μl), but not neurotrophin-3 injection (1 µg/50 µl), resulted in long-lasting facial heat hyperalgesia, which was both significantly reduced by previous neurotrophin-3 local treatment. In conclusion, we suggest that neurotrophin-3 is a potent modulator of facial heat hyperalgesia, which may exert an inhibitory influence on the trkA pathway. Neurotrophin-3 treatment may represent a promising approach, especially in pain conditions associated with increased levels of nerve growth factor.
Sigma-Aldrich
Human NT-3 ELISA Kit, for serum, plasma, cell culture supernatant and urine