Merck
  • Assessment of cadmium and lead contamination in rice farming soils and rice (Oryza sativa L.) from Guayas province in Ecuador.
Assessment of cadmium and lead contamination in rice farming soils and rice (Oryza sativa L.) from Guayas province in Ecuador.
Environmental pollution (Barking, Essex : 1987) (2020-02-12)
Martin Ochoa, Wladimir Tierra, Diego Santiago Tupuna-Yerovi, Danilo Guanoluisa, Xosé Luis Otero, Jenny Ruales
ABSTRACT
Rice is the world's most consumed and in-demand grain. Ecuador is one of the main rice-consuming countries in Latin America, with an average per capita consumption of 53.2 kg per year. Rice cultivation takes place under flooding conditions, which favors the mobilization and subsequent accumulation of heavy metals in the plant. This study's principal objective was to evaluate the contamination of cadmium (Cd) and lead (Pb) in the rice cultivation system in the province of Guayas. To this end, extensive sampling of water, soil and rice grains was carried. Water samples were analyzed to determine physicochemical properties and concentrations of dissolved Cd and Pb. Physicochemical properties, total organic carbon (TOC), total content of nitrogen (N), iron (Fe), manganese (Mn), phosphorus (P), bioavailable phosphorus (P mehlich), Cd and Pb were determined in soil samples. In addition, to understand the dynamics of Cd and Pb mobility and bioavailability, an extraction of six randomly selected soil samples was carried out. The concentration values of the total Cd and Pb content in the rice cultivation system did not exceed the maximum recommended limit for soil, water and rice grains. However, 85% of the total Cd was in the soluble or exchangeable fraction of the soil, while the Pb was strongly bound to crystalline iron oxyhydroxides. It was established that the TOC, N, Fe, and P mehlich have a significant correlation (p < 0.05) with the overall concentration of Cd and Pb in the rice farming soil. The Cd and Pb present in rice do not represent a dietary health risk to the population of Ecuador.
Product Description
Supelco
EPN, PESTANAL®, analytical standard