- Competitive and selective antagonism of P2Y1 receptors by N6-methyl 2'-deoxyadenosine 3',5'-bisphosphate.
Competitive and selective antagonism of P2Y1 receptors by N6-methyl 2'-deoxyadenosine 3',5'-bisphosphate.
British journal of pharmacology (1998-06-18)
J L Boyer, A Mohanram, E Camaioni, K A Jacobson, T K Harden
PMID9630335
ABSTRACT
The antagonist activity of N6-methyl 2'-deoxyadenosine 3',5'-bisphosphate (N6MABP) has been examined at the phospholipase C-coupled P2Y1 receptor of turkey erythrocyte membranes. N6MABP antagonized 2MeSATP-stimulated inositol phosphate hydrolysis with a potency approximately 20 fold greater than the previously studied parent molecule, adenosine 3',5'-bisphosphate. The P2Y1 receptor antagonism observed with N6MABP was competitive as revealed by Schild analysis (pK(B) = 6.99 +/- 0.13). Whereas N6MABP was an antagonist at the human P2Y1 receptor, no antagonist effect of N6MABP was observed at the human P2Y2, human P2Y4 or rat P2Y6 receptors.
MATERIALS