Merck
  • A sex difference in the response of the rodent postsynaptic density to synGAP haploinsufficiency.
A sex difference in the response of the rodent postsynaptic density to synGAP haploinsufficiency.
eLife (2020-01-16)
Tara L Mastro, Anthony Preza, Shinjini Basu, Sumantra Chattarji, Sally M Till, Peter C Kind, Mary B Kennedy
ABSTRACT
SynGAP is a postsynaptic density (PSD) protein that binds to PDZ domains of the scaffold protein PSD-95. We previously reported that heterozygous deletion of Syngap1 in mice is correlated with increased steady-state levels of other key PSD proteins that bind PSD-95, although the level of PSD-95 remains constant (Walkup et al., 2016). For example, the ratio to PSD-95 of Transmembrane AMPA-Receptor-associated Proteins (TARPs), which mediate binding of AMPA-type glutamate receptors to PSD-95, was increased in young Syngap1+/-mice. Here we show that only females and not males show a highly significant correlation between an increase in TARP and a decrease in synGAP in the PSDs of Syngap1+/-rodents. The data reveal a sex difference in the adaptation of the PSD scaffold to synGAP haploinsufficiency.
Sigma-Aldrich
CRISPR/Cas9 Products and Services, Design and order CRISPR gRNA, Cas9, screening libraries, controls and companion products. Formats include plant, lentivirus, IVT-RNA, plasmid, synthetic, and protein.
Sigma-Aldrich
Anti-Stargazin Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Phenylmethanesulfonyl chloride, 98%