Merck
  • Isolation and characterization of violacein from an Antarctic Iodobacter: a non-pathogenic psychrotolerant microorganism.
Isolation and characterization of violacein from an Antarctic Iodobacter: a non-pathogenic psychrotolerant microorganism.
Extremophiles : life under extreme conditions (2019-07-22)
Joaquin Atalah, Lotsé Blamey, Sebastian Muñoz-Ibacache, Felipe Gutierrez, Marcela Urzua, Maria Victoria Encinas, Maritza Páez, Junsong Sun, Jenny M Blamey
ABSTRACT
Violacein is an intensely purple pigment synthesized by various genera of bacteria that has been discovered to have a wide range of interesting biological activities which range from anticarcinogenic to antibacterial. One of the hindrances for its real-life application is that the first microorganisms found to produce the compound may act as opportunistic pathogens. Here, we report the isolation and characterization of violacein from a non-pathogenic Antarctic Iodobacter strain. Its anti-microbial properties were also tested. The method proposed here for the purification of violacein shows high yields, indicating that this Antarctic microorganism could be a valuable source for this important pigment. This is the first characterization of violacein from an Antarctic Iodobacter strain and here we also present a viable method to obtain this pigment for potential biotechnological applications.
Sigma-Aldrich
Violacein from Janthinobacterium lividum, >98% (violacein (minimum 85% violacein) and deoxyviolacein, HPLC)