Merck
  • CD73 Promotes Glioblastoma Pathogenesis and Enhances Its Chemoresistance via A2B Adenosine Receptor Signaling.
CD73 Promotes Glioblastoma Pathogenesis and Enhances Its Chemoresistance via A2B Adenosine Receptor Signaling.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2019-03-31)
Angela Yan, Michelle L Joachims, Linda F Thompson, Andrew D Miller, Peter D Canoll, Margaret S Bynoe
ABSTRACT
Glioblastoma (GB) is one of the deadliest brain cancers to afflict humans, and it has a very poor survival rate even with treatment. The extracellular adenosine-generating enzyme CD73 is involved in many cellular functions that can be usurped by tumors, including cell adhesion, proliferation, invasion, and angiogenesis. We set out to determine the role of CD73 in GB pathogenesis. To do this, we established a unique GB mouse model (CD73-FLK) in which we spatially expressed CD73 on endothelial cells in CD73-/- mice. This allowed us to elucidate the mechanism of host CD73 versus GB-expressed CD73 by comparing GB pathogenesis in WT, CD73-/-, and CD73-FLK mice. GB in CD73-/- mice had decreased tumor size, decreased tumor vessel density, and reduced tumor invasiveness compared with GB in WT mice. Interestingly, GBs in CD73-FLK mice were much more invasive and caused complete distortion of the brain morphology. We showed a 20-fold upregulation of A2B AR on GB compared with sham, and its activation induced matrix metalloproteinase-2, which enhanced GB pathogenesis. Inhibition of A2B AR signaling decreased multidrug resistance transporter protein expression, including permeability glycoprotein (P-gp) and multidrug resistance-associated protein 1 (MRP1). Further, we showed that blockade of A2B AR signaling potently increased GB cell death induced by the chemotherapeutic drug temozolomide. Together, these findings suggest that CD73 and A2B AR play a multifaceted role in GB pathogenesis and progression and that targeting the CD73-A2B AR axis can benefit GB patients and inform new approaches for therapy to treat GB patients.SIGNIFICANCE STATEMENT Glioblastoma (GB) is the most devastating primary brain tumor. GB patients' median survival is 16 months even with treatment. It is critical that we develop prophylaxes to advance GB treatment and improve patient survival. CD73-generated adenosine has been implicated in cancer pathogenesis, but its role in GB was not ascertained. Here, we demonstrated that host CD73 plays a prominent role in multiple areas of glioblastoma pathogenesis, including promoting GB growth, its angiogenesis, and its invasiveness. We found a 20-fold increase in A2B adenosine receptor (AR) expression on GB compared with sham, and its inhibition increased GB chemosensitivity to temozolomide. These findings strongly indicate that blockade or inhibition of CD73 and the A2B AR are prime targets for future GB therapy.
Sigma-Aldrich
Anti-MMP-2 Antibody, a.a. 468-483 hMMP2, clone 42-5D11, clone 42-5D11, Chemicon®, from mouse
Sigma-Aldrich
Anti-α-Dystroglycan Antibody, clone VIA4-1, culture supernatant, clone VIA4-1, Upstate®
Sigma-Aldrich
Anti-MMP-9 Antibody, Catalytic domain, Chemicon®, from rabbit