Merck
Identifying the murine mammary cell target of metformin exposure.
Communications biology (2019-05-28)
Mona Shehata, Hyeyeon Kim, Ravi Vellanki, Paul D Waterhouse, Mathepan Mahendralingam, Alison E Casey, Marianne Koritzinsky, Rama Khokha
ABSTRACT
The heterogeneity of breast cancer makes current therapies challenging. Metformin, the anti-diabetic drug, has shown promising anti-cancer activities in epidemiological studies and breast cancer models. Yet, how metformin alters the normal adult breast tissue remains elusive. We demonstrate metformin intake at a clinically relevant dose impacts the hormone receptor positive (HR+) luminal cells in the normal murine mammary gland. Metformin decreases total cell number, progenitor capacity and specifically reduces DNA damage in normal HR+ luminal cells, decreases oxygen consumption rate and increases cell cycle length of luminal cells. HR+ luminal cells demonstrate the lowest levels of mitochondrial respiration and capacity to handle oxidative stress compared to the other fractions, suggesting their intrinsic susceptibility to long-term metformin exposure. Uncovering HR+ luminal cells in the normal mammary gland as the major cell target of metformin exposure could identify patients that would most benefit from repurposing this anti-diabetic drug for cancer prevention/therapy purposes.
Sigma-Aldrich
Anti-ALDH1A3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution