Merck
  • Phospholipase A2 activity-dependent stimulation of Ca2+ entry by human parvovirus B19 capsid protein VP1.
Phospholipase A2 activity-dependent stimulation of Ca2+ entry by human parvovirus B19 capsid protein VP1.
Journal of virology (2006-09-08)
Adrian Lupescu, C-Thomas Bock, Philipp A Lang, Susanne Aberle, Heike Kaiser, Reinhard Kandolf, Florian Lang
ABSTRACT
Recent reports demonstrated an association of human parvovirus B19 with inflammatory cardiomyopathy (iCMP), which is accompanied by endothelial dysfunction. As intracellular Ca(2+) activity is a key regulator of cell function and participates in mechanisms leading to endothelial dysfunction, the present experiments explored the effects of the B19 capsid proteins VP1 and VP2. A secreted phospholipase A2 (PLA2)-like activity has been located in the VP1 unique region of the B19 minor capsid protein. As PLA2 has recently been shown to activate the store-operated or capacitative Ca(2+) channel I(CRAC), we analyzed the impact of the viral PLA2 motif on Ca(2+) entry. We cloned the VP1 and VP2 genes isolated from a patient suffering from fatal B19 iCMP into eukaryotic expression vectors. We also generated a B19 replication-competent plasmid to demonstrate PLA2 activity under the control of the complete B19 genome. After the transfection of human endothelial cells (HMEC-1), cytosolic Ca(2+) activity was determined by utilizing Fura-2 fluorescence. VP1 and VP2 expression did not significantly modify basal cytosolic Ca(2+) activity or the decline of cytosolic Ca(2+) activity following the removal of extracellular Ca(2+). However, expression of VP1 and of the full-length B19 clone, but not of VP2, significantly accelerated the increase of cytosolic Ca(2+) activity following the readdition of extracellular Ca(2+) in the presence of thapsigargin, indicating an activation of I(CRAC.) The effect of VP1 was mimicked by the PLA2 product lysophosphatidylcholine and abolished by an inactivating mutation of the PLA2-encoding region of the VP1 gene. Our observations point to the activation of Ca(2+) entry by VP1 PLA2 activity, an effect likely participating in the pathophysiology of B19 infection.
Sigma-Aldrich
Anti-Parvovirus B19 Antibody, aa 328-344 of VP2 capsid protein, clone R92F6, clone R92F6, Chemicon®, from mouse
Sigma-Aldrich
Anti-Parvovirus B19 Antibody, clone 521-5D, clone 521-5D, Chemicon®, from mouse