Merck
  • Deletion of CD38 Suppresses Glial Activation and Neuroinflammation in a Mouse Model of Demyelination.
Deletion of CD38 Suppresses Glial Activation and Neuroinflammation in a Mouse Model of Demyelination.
Frontiers in cellular neuroscience (2019-06-28)
Jureepon Roboon, Tsuyoshi Hattori, Hiroshi Ishii, Mika Takarada-Iemata, Thuong Manh Le, Yoshitake Shiraishi, Noriyuki Ozaki, Yasuhiko Yamamoto, Akira Sugawara, Hiroshi Okamoto, Haruhiro Higashida, Yasuko Kitao, Osamu Hori
ABSTRACT
CD38 is an enzyme that catalyzes the synthesis of cyclic adenosine diphosphate-ribose from nicotinamide adenine dinucleotide (NAD+). We recently reported that this molecule regulates the maturation and differentiation of glial cells such as astrocytes and oligodendrocytes (OLs) in the developing brain. To analyze its role in the demyelinating situation, we employed cuprizone (CPZ)-induced demyelination model in mice, which is characterized by oligodendrocyte-specific apoptosis, followed by the strong glial activation, demyelination, and repopulation of OLs. By using this model, we found that CD38 was upregulated in both astrocytes and microglia after CPZ administration. Experiments using wild-type and CD38 knockout (KO) mice, together with those using cultured glial cells, revealed that CD38 deficiency did not affect the initial decrease of the number of OLs, while it attenuated CPZ-induced demyelination, and neurodegeneration. Importantly, the clearance of the degraded myelin and oligodendrocyte repopulation were also reduced in CD38 KO mice. Further experiments revealed that these observations were associated with reduced levels of glial activation and inflammatory responses including phagocytosis, most likely through the enhanced level of NAD+ in CD38-deleted condition. Our results suggest that CD38 and NAD+ in the glial cells play a critical role in the demyelination and subsequent oligodendrocyte remodeling through the modulation of glial activity and neuroinflammation.
Sigma-Aldrich
Anti-Myelin Basic Protein Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-APP A4 Antibody, a.a. 66-81 of APP {NT}, clone 22C11, clone 22C11, Chemicon®, from mouse
Sigma-Aldrich
Anti-Glutamate Receptor 2 & 4 Antibody, clone 3A11, clone 3A11, Chemicon®, from mouse