Merck
  • A novel strategy to inhibit FAK and IGF-1R decreases growth of pancreatic cancer xenografts.
A novel strategy to inhibit FAK and IGF-1R decreases growth of pancreatic cancer xenografts.
Molecular carcinogenesis (2009-11-04)
Donghang Zheng, Vita Golubovskaya, Elena Kurenova, Cheng Wood, Nicole A Massoll, David Ostrov, William G Cance, Steven N Hochwald
ABSTRACT
Deregulation of insulin-like growth factor-1 receptor (IGF-1R) and focal adhesion kinase (FAK) signaling pathways plays an important role in cancer cell proliferation and metastasis. In pancreatic cancer cells, the crosstalk and compensatory mechanisms between these two pathways reduce the efficacy of the treatments that target only one of the pathways. Ablation of IGF-1R signaling by siRNA showed minimal effects on the survival and growth of pancreatic cancer cells. An increased activity of FAK pathway was seen in these cells after IGF-1R knockdown. Further inhibition of FAK pathway using Y15 significantly decreased cell survival, adhesion, and promoted apoptosis. The combination of Y15 treatment and IGF-1R knockdown also showed significant antitumor effect in vivo. The current study demonstrates the importance of dual inhibition of both these signaling pathways as a novel strategy to decrease both in vitro and in vivo growth of human pancreatic cancer.
Sigma-Aldrich
Anti-FAK Antibody, clone 4.47, clone 4.47, Upstate®, from mouse
Sigma-Aldrich
Anti-IGF-IR (Ab-1) Mouse mAb (αIR3), liquid, clone αIR3, Calbiochem®