Merck
  • Evidence for functional pre-coupled complexes of receptor heteromers and adenylyl cyclase.
Evidence for functional pre-coupled complexes of receptor heteromers and adenylyl cyclase.
Nature communications (2018-03-30)
Gemma Navarro, Arnau Cordomí, Verónica Casadó-Anguera, Estefanía Moreno, Ning-Sheng Cai, Antoni Cortés, Enric I Canela, Carmen W Dessauer, Vicent Casadó, Leonardo Pardo, Carme Lluís, Sergi Ferré
ABSTRACT
G protein-coupled receptors (GPCRs), G proteins and adenylyl cyclase (AC) comprise one of the most studied transmembrane cell signaling pathways. However, it is unknown whether the ligand-dependent interactions between these signaling molecules are based on random collisions or the rearrangement of pre-coupled elements in a macromolecular complex. Furthermore, it remains controversial whether a GPCR homodimer coupled to a single heterotrimeric G protein constitutes a common functional unit. Using a peptide-based approach, we here report evidence for the existence of functional pre-coupled complexes of heteromers of adenosine A2A receptor and dopamine D2 receptor homodimers coupled to their cognate Gs and Gi proteins and to subtype 5 AC. We also demonstrate that this macromolecular complex provides the necessary frame for the canonical Gs-Gi interactions at the AC level, sustaining the ability of a Gi-coupled GPCR to counteract AC activation mediated by a Gs-coupled GPCR.
Sigma-Aldrich
Anti-Adenosine Receptor A2a Antibody, clone 7F6-G5-A2, clone 7F6-G5-A2, Upstate®, from mouse
Sigma-Aldrich
Anti-Adenosine A2a Receptor Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Dopamine D2 Receptor Antibody, Chemicon®, from rabbit