- Direct Synthesis of N-Alkyl Arylglycines by Organocatalytic Asymmetric Transfer Hydrogenation of N-Alkyl Aryl Imino Esters.
Direct Synthesis of N-Alkyl Arylglycines by Organocatalytic Asymmetric Transfer Hydrogenation of N-Alkyl Aryl Imino Esters.
Organic letters (2017-10-06)
Javier Mazuela, Thomas Antonsson, Magnus J Johansson, Laurent Knerr, Stephen P Marsden
PMID28981292
ABSTRACT
The organocatalytic asymmetric transfer hydrogenation of N-alkyl aryl imino esters for the direct synthesis of N-alkylated arylglycinate esters is reported. High yields and enantiomeric ratios were obtained, and tolerance to a diverse set of functional groups facilitated the preparation of more complex molecules as well as intermediates for active pharmaceuticals. A simple recycling protocol was developed for the Brønsted acid catalyst which could be reused through five cycles with no loss of activity or selectivity.