- Antifungal Properties of Two Volatile Organic Compounds on Barley Pathogens and Introduction to Their Mechanism of Action.
Antifungal Properties of Two Volatile Organic Compounds on Barley Pathogens and Introduction to Their Mechanism of Action.
International journal of environmental research and public health (2019-08-14)
Amine Kaddes, Marie-Laure Fauconnier, Khaled Sassi, Bouzid Nasraoui, M Haïssam Jijakli
PMID31405165
ABSTRACT
This study evaluated the antifungal effects of various volatile organic compounds (VOCs) against two common pathogens: Fusarium culmorum and Cochliobolus sativus. Among the various VOCs, methyl propanoate (MP) and methyl prop-2-enoate (MA) exhibited remarkable antifungal effects under different experimental conditions (direct or indirect contact) and at different concentrations (500-1000 μM). In addition, the type of antifungal effect (fungistatic or fungicidal) appeared to be strongly correlated with the VOC concentrations. Additional tests revealed that both molecules increased membrane permeability of pathogenic spores, which resulted in a decreased efflux of K+ ions into the intracellular medium.