Merck
  • A Green Water-Soluble Cyclophosphazene as a Flame Retardant Finish for Textiles.
A Green Water-Soluble Cyclophosphazene as a Flame Retardant Finish for Textiles.
Molecules (Basel, Switzerland) (2019-08-29)
Thomas Mayer-Gall, Dennis Plohl, Leonie Derksen, Dana Lauer, Pia Neldner, Wael Ali, Sabine Fuchs, Jochen S Gutmann, Klaus Opwis
ABSTRACT
Poly- and cyclophosphazenes are excellent flame retardants but currently, are not used as textile finishing agents because water-soluble and permanent washing systems are missing. Here, we demonstrate for the first time, the successful usage of a water-soluble cyclotriphosphazene derivative for textile finishing for cotton, different cotton/polyester, and cotton/polyamide blend fabrics. A durable finish was achieved using a photoinduced grafting reaction. The flame retardant properties of the various fabrics were improved with a higher limiting oxygen index, a reduced heat release rate, and an exhibition of intumescent. Furthermore, the finished textiles passed several standardized flammability tests.