Merck
  • Leptomycin B targets a regulatory cascade of crm1, a fission yeast nuclear protein, involved in control of higher order chromosome structure and gene expression.
Leptomycin B targets a regulatory cascade of crm1, a fission yeast nuclear protein, involved in control of higher order chromosome structure and gene expression.
The Journal of biological chemistry (1994-03-04)
K Nishi, M Yoshida, D Fujiwara, M Nishikawa, S Horinouchi, T Beppu
ABSTRACT
The molecular action of leptomycin B (LMB), an agent inducing arrest of the eukaryotic cell cycle at G1 and G2 phases, was investigated by analyzing an LMB resistance gene of Schizosaccharomyces pombe. A genomic library of an LMB-resistant mutant was screened for LMB resistance, and a DNA fragment containing an open reading frame (ORF) of 1078 amino acids was cloned on a multicopy vector. The plasmid was found to confer drug resistance specifically to LMB. Nucleotide sequencing revealed that the ORF was a mutant gene for the essential nuclear protein crm1, which had been reported to complement a cold-sensitive mutation causing deformed nuclear morphology. The gene product named crm1-N1 had two amino acid replacements (Gly-503 to Asp and Met-546 to Ile). Two allelic mutants of crm1 (crm1-809 and crm1-119) were found to be hypersensitive and resistant, respectively, to LMB. Nuclear morphology of the cold-sensitive crm1-809 mutant at the restrictive temperature was almost the same as that of the wild-type cells treated with LMB. Furthermore, a low concentration of LMB induced the intracellular accumulation of a 25-kDa protein in the wild-type cells, which was immunologically identical to the protein accumulating in the crm1-809 mutant cells. These results strongly suggest that LMB primarily inhibits the function of the crm1 gene which is required for maintaining higher order chromosome structures, correct gene expression, and cell growth in the fission yeast.
Sigma-Aldrich
Leptomycin B solution from Streptomyces sp., ≥95% (HPLC), Supplied in methanol: water (7:3)