- The ORMDL3 Asthma Gene Regulates ICAM1 and Has Multiple Effects on Cellular Inflammation.
The ORMDL3 Asthma Gene Regulates ICAM1 and Has Multiple Effects on Cellular Inflammation.
Polymorphisms on chromosome 17q21 confer the major genetic susceptibility to childhood-onset asthma. Risk alleles positively correlate with ORMDL3 (orosomucoid-like 3) expression. The locus influences disease severity and the frequency of human rhinovirus (HRV)-initiated exacerbations. ORMDL3 is known to regulate sphingolipid synthesis by binding serine palmitoyltransferase, but its role in inflammation is incompletely understood. To investigate the role of ORMDL3 in cellular inflammation. We modeled a time series of IL1B-induced inflammation in A549 cells, using cytokine production as outputs and testing effects of ORMDL3 siRNA knockdown, ORMDL3 overexpression, and the serine palmitoyltransferase inhibitor myriocin. We replicated selected findings in normal human bronchial epithelial cells. Cytokine and metabolite levels were analyzed by analysis of variance. Transcript abundances were analyzed by group means parameterization, controlling the false discovery rate below 0.05. Silencing ORMDL3 led to steroid-independent reduction of IL6 and IL8 release and reduced endoplasmic reticulum stress after IL1B stimulation. Overexpression and myriocin conversely augmented cytokine release. Knockdown reduced expression of genes regulating host-pathogen interactions, stress responses, and ubiquitination: in particular, ORMDL3 knockdown strongly reduced expression of the HRV receptor ICAM1. Silencing led to changes in levels of transcripts and metabolites integral to glycolysis. Increased levels of ceramides and the immune mediator sphingosine-1-phosphate were also observed. The results show ORMDL3 has pleiotropic effects during cellular inflammation, consistent with its substantial genetic influence on childhood asthma. Actions on ICAM1 provide a mechanism for the locus to confer susceptibility to HRV-induced asthma.